Gene Transcripts
- Gene Symbol
- Cdc25b
- Entrez Gene ID
- 12531
- Gene Name
- cell division cycle 25B
- Gene Aliases
- AI604853
- Chromosome Location
- Chr. 2: 131186436 - 131198511 on Build GRCm38
- UniGene ID
- Mm.38444
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111075.4 |
NP_001104545.1
|
3, 4 |
615 |
|
NM_023117.4 |
NP_075606.1
|
3, 4 |
615 |
|
GenBank mRNA |
AK004836.1 |
- |
3, 4 |
572 |
|
AK043730.1 |
- |
3, 4 |
606 |
|
AK153903.1 |
- |
3, 4 |
616 |
|
AK157397.1 |
- |
3, 4 |
573 |
|
BC048164.1 |
- |
3, 4 |
612 |
|
BC057568.1 |
- |
3, 4 |
486 |
6401489 |
BQ561430.1 |
- |
3, 4 |
271 |
|
S93521.1 |
- |
3, 4 |
484 |
|
View Full Product Details