Gene Transcripts

Gene Symbol
H1F0
Entrez Gene ID
3005
Gene Name
H1 histone family member 0
Gene Aliases
H10, H1FV
Chromosome Location
Chr. 22: 37805107 - 37807436 on Build GRCh38
UniGene ID
Hs.745024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK091372.1 - 2 1318
BC000145.1 - 1 1374 3049108
BC029046.1 - 1 1373 5189077

View Full Product Details