Gene Transcripts

Gene Symbol
Trim47
Entrez Gene ID
217333
Gene Name
tripartite motif-containing 47
Gene Aliases
2210023F24Rik
Chromosome Location
Chr. 11: 116105750 - 116110235 on Build GRCm38
UniGene ID
Mm.347770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205081.1 NP_001192010.1 6 1911
NM_172570.5 NP_766158.3 6 1908
GenBank mRNA AK031572.1 - 6 1883
AK075838.1 - 6 1337
BC023393.1 - 4 1083
BC024468.1 - 4 952
BC043714.1 - 5 1223
BC055038.1 - 4 1208
BC064728.1 - 6 1305
BC080731.1 - 6 1307
BC100317.1 - 6 1892
BC132148.1 - 6 1689 40130425
BC137953.1 - 6 1689 8860972
BC145059.1 - 6 1690 9053607

View Full Product Details