Gene Transcripts

Gene Symbol
Adck2
Entrez Gene ID
57869
Gene Name
aarF domain containing kinase 2
Gene Aliases
AA408112, BB006057
Chromosome Location
Chr. 6: 39573835 - 39588769 on Build GRCm38
UniGene ID
Mm.247957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178873.3 NP_849204.1 1 655
GenBank mRNA AK028557.1 - 1 659
AK028568.1 - 1 656
AK161225.1 - 1 658
BC069944.1 - 1 403 30446117

View Full Product Details