Gene Transcripts

Gene Symbol
Sdad1
Entrez Gene ID
231452
Gene Name
SDA1 domain containing 1
Gene Aliases
4931421J16, AA591032, AW538460
Chromosome Location
Chr. 5: 92284010 - 92310024 on Build GRCm38
UniGene ID
Mm.86564
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172713.2 NP_766301.1 3 415
GenBank mRNA AK029881.1 - 3 417
AK160646.1 - 3 286
AK164608.1 - 3 381
BC042708.1 - 3 365 5024980

View Full Product Details