Gene Transcripts

Gene Symbol
Banf2
Entrez Gene ID
403171
Gene Name
barrier to autointegration factor 2
Gene Aliases
4930517K23Rik, Baf-L, Baf-like, Gm115
Chromosome Location
Chr. 2: 144033102 - 144073979 on Build GRCm38
UniGene ID
Mm.389362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044750.1 NP_001038215.1 2 161
NM_207275.1 NP_997158.1 3 281
GenBank mRNA AK076863.1 - 3 281
BC049628.1 - 161 6706626

View Full Product Details