Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025163.1 | NP_001020334.1 | 5 | 588 | |
NM_001112805.1 | NP_001106276.1 | 4 | 492 | ||
NM_177888.4 | NP_808556.1 | 5 | 731 | ||
GenBank mRNA | AK039658.1 | - | 5 | 704 | |
AK134974.1 | - | 5 | 617 | ||
AK163133.1 | - | 4 | 491 | ||
AK173164.1 | - | 4 | 512 | ||
BC085322.1 | - | 5 | 579 | 30739758 | |
U29511.1 | - | 2 | 280 |