Gene Transcripts

Gene Symbol
Ap1ar
Entrez Gene ID
211556
Gene Name
adaptor-related protein complex 1 associated regulatory protein
Gene Aliases
AA407621, C78909, Gadkin
Chromosome Location
Chr. 3: 127807007 - 127837518 on Build GRCm38
UniGene ID
Mm.403486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145964.2 NP_666076.2 10 1628
GenBank mRNA BC002199.1 - 1 838

View Full Product Details