Gene Transcripts

Gene Symbol
ALYREF
Entrez Gene ID
10189
Gene Name
Aly/REF export factor
Gene Aliases
ALY, ALY/REF, BEF, REF, THOC4
Chromosome Location
Chr. 17: 81887835 - 81891622 on Build GRCh38
UniGene ID
Hs.534385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005782.3 NP_005773.3 3 518
GenBank mRNA AF047002.1 - 3 419
AF086513.1 - 2 173
BC052302.1 - 3 518 6381528

View Full Product Details