Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172609.3 | NP_766197.2 | 3, 4 | 377 | |
GenBank mRNA | AK008133.1 | - | 3, 4 | 380 | |
AK009868.1 | - | 3, 4 | 377 | ||
AK013471.1 | - | 3, 4 | 366 | ||
AK049442.1 | - | 365 | |||
AK150360.1 | - | 3, 4 | 365 | ||
AK150787.1 | - | 3, 4 | 374 | ||
AK152843.1 | - | 3, 4 | 365 | ||
AK167803.1 | - | 3, 4 | 365 | ||
BC056920.1 | - | 3, 4 | 359 | 6819748 |