Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277305.1 | NP_001264234.1 | 2 | 420 | |
NM_175643.3 | NP_783574.1 | 2 | 420 | ||
GenBank mRNA | AK220464.1 | - | 1 | 162 | |
BC046456.1 | - | 2 | 420 | 3965027 | |
CJ068037.1 | - | 2 | 416 |