Gene Transcripts

Gene Symbol
Ripply3
Entrez Gene ID
170765
Gene Name
ripply3 homolog (zebrafish)
Gene Aliases
Dscr6
Chromosome Location
Chr. 16: 94328422 - 94336935 on Build GRCm38
UniGene ID
Mm.42604
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133229.2 NP_573492.2 4 1493
GenBank mRNA AB063284.1 - 4 1492
AK041355.1 - 4 1483
AK172430.1 - 4 1483

View Full Product Details