Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172438.3 | NP_766026.1 | 5 | 451 | |
GenBank mRNA | AK050734.1 | - | 5 | 451 | |
AK173078.1 | - | 3 | 289 | ||
AK207100.1 | - | 2 | 117 | ||
BC020332.1 | - | 4 | 416 | ||
BC021779.1 | - | 4 | 425 | ||
BC039758.1 | - | 5 | 452 | 3995737 |