Gene Transcripts

Gene Symbol
Ric8
Entrez Gene ID
101489
Gene Name
resistance to inhibitors of cholinesterase 8 homolog (C. elegans)
Gene Aliases
AI114950, RIC-8, Ric8a
Chromosome Location
Chr. 7: 140857397 - 140863731 on Build GRCm38
UniGene ID
Mm.281916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053194.4 NP_444424.1 10 2269
GenBank mRNA AF288813.1 - 10 2268
AK031466.1 - 10 2216
AK165536.1 - 10 2246
AK167746.1 - 10 2272
AK169367.1 - 10 2233
AK210129.1 - 1 78
BC005633.1 - 2 2041
BC022917.1 - 10 2246 4460521

View Full Product Details