Gene Transcripts

Gene Symbol
Spinkl
Entrez Gene ID
77424
Gene Name
serine protease inhibitor, Kazal type-like
Gene Aliases
9530002K18Rik
Chromosome Location
Chr. 18: 44166358 - 44176707 on Build GRCm38
UniGene ID
Mm.99224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183123.2 NP_898946.1 3, 4 170
GenBank mRNA AK020535.1 - 3, 4 171
EU234015.1 - 548

View Full Product Details