Gene Transcripts

Gene Symbol
Poteg
Entrez Gene ID
70952
Gene Name
POTE ankyrin domain family, member G
Gene Aliases
4921537P18Rik, 4930456F22Rik, Gsarp1, Gsarp2
Chromosome Location
Chr. 8: 27447529 - 27582715 on Build GRCm38
UniGene ID
Mm.158160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026256.2 NP_080532.1 9 1241
GenBank mRNA AK015165.1 - 9 1242
AK015473.1 - 9 1242
DQ298444.1 - 9 1242

View Full Product Details