Gene Transcripts

Gene Symbol
Scara5
Entrez Gene ID
71145
Gene Name
scavenger receptor class A, member 5 (putative)
Gene Aliases
4932433F15Rik, 4933425F03Rik, AV278087, Tesr
Chromosome Location
Chr. 14: 65666403 - 65764826 on Build GRCm38
UniGene ID
Mm.83840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028903.2 NP_083179.2 9 2301
GenBank mRNA AK084591.1 - 128
AK133176.1 - 9 2302
BC016096.1 - 9 2277 4506472
BC023907.1 - 9 2217 5321978
BC026758.1 - 6 1322
BC032159.1 - 9 2257 5354657

View Full Product Details