Gene Transcripts

Gene Symbol
Spp2
Entrez Gene ID
75396
Gene Name
secreted phosphoprotein 2
Gene Aliases
0610038O04Rik, pp-24, spp24
Chromosome Location
Chr. 1: 88406967 - 88428300 on Build GRCm38
UniGene ID
Mm.173058
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029269.2 NP_083545.1 6, 7 655
GenBank mRNA AJ315513.1 - 6, 7 616
AK002814.1 - 6, 7 656
BC027494.1 - 6, 7 581 1433655

View Full Product Details