Gene Transcripts

Gene Symbol
Mrps9
Entrez Gene ID
69527
Gene Name
mitochondrial ribosomal protein S9
Gene Aliases
2310002A08Rik, AW322516, Mrp-S9, S9mt, mg637
Chromosome Location
Chr. 1: 42851233 - 42905683 on Build GRCm38
UniGene ID
Mm.252982
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023514.4 NP_076003.3 1, 2 202
GenBank mRNA AK009082.1 - 1, 2 152
AK031497.1 - 1, 2 151
AK049465.1 - 1, 2 160
AK144656.1 - 1, 2 160
AK150985.1 - 1, 2 160
AK214977.1 - 1, 2 40
BC116437.2 - 1, 2 152 40104493
BY145802.1 - 1, 2 202

View Full Product Details