Gene Transcripts

Gene Symbol
Bbs2
Entrez Gene ID
67378
Gene Name
Bardet-Biedl syndrome 2 (human)
Gene Aliases
2410125H22Rik, AI447581
Chromosome Location
Chr. 8: 94067954 - 94099370 on Build GRCm38
UniGene ID
Mm.292107
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026116.2 NP_080392.1 11 1727
GenBank mRNA AF342737.1 - 11 1727
AK010779.1 - 11 1340
BC057184.1 - 11 1386 5066846

View Full Product Details