Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026210.4 | NP_080486.1 | 7 | 1170 | |
GenBank mRNA | AK007707.1 | - | 7 | 1086 | |
AK017572.1 | - | 7 | 1084 | ||
AK050027.1 | - | 7 | 1206 | ||
AK077277.1 | - | 7 | 1172 | ||
AK150936.1 | - | 7 | 1162 |