Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026369.2 | NP_080645.2 | 4 | 1439 | |
GenBank mRNA | AK008097.1 | - | 4 | 1329 | |
AK017669.1 | - | 4 | 1317 | ||
AK017985.1 | - | 4 | 1346 | ||
AK087427.1 | - | 4 | 1306 | ||
AK149756.1 | - | 4 | 1308 | ||
AK150443.1 | - | 4 | 1316 | ||
AK150703.1 | - | 4 | 1316 | ||
AK151490.1 | - | 3 | 1124 | ||
AK151654.1 | - | 4 | 1307 | ||
AK151656.1 | - | 4 | 1330 | ||
AK152281.1 | - | 4 | 1330 | ||
AK153262.1 | - | 4 | 1310 | ||
AK159407.1 | - | 4 | 1363 | ||
AK159438.1 | - | 4 | 1303 | ||
AK159738.1 | - | 4 | 1326 | ||
AK166284.1 | - | 4 | 1333 | ||
AK166335.1 | - | 4 | 1347 | ||
AK166725.1 | - | 4 | 1330 | ||
AK166919.1 | - | 4 | 1332 | ||
AK167144.1 | - | 3 | 1086 | ||
AK168264.1 | - | 4 | 1307 | ||
AK168986.1 | - | 4 | 1278 | ||
BC060143.1 | - | 4 | 1345 | 6411792 |