Gene Transcripts

Gene Symbol
Carhsp1
Entrez Gene ID
52502
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
1200011K09Rik, Crhsp-24, D16Ertd465e
Chromosome Location
Chr. 16: 8658580 - 8672153 on Build GRCm38
UniGene ID
Mm.142095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025821.2 NP_080097.1 4 382
GenBank mRNA AF414101.1 - 3 334
AK002853.1 - 4 383
AK003926.1 - 4 385
AK004711.2 - 4 382
AK143612.1 - 4 383
AK149875.1 - 4 386
AK150245.1 - 4 363
AK154460.1 - 4 383
AK155318.1 - 4 385
AK158112.1 - 4 386
AK158252.1 - 4 385
AK165373.1 - 4 385
AK168504.1 - 4 383
AK170411.1 - 4 380
AK171282.1 - 4 385
AK203087.1 - 3 194
BC011225.1 - 3 352 4166353
BC086754.1 - 4 365 6812962

View Full Product Details