Gene Transcripts
- Gene Symbol
- Atg13
- Entrez Gene ID
- 51897
- Gene Name
- autophagy related 13
- Gene Aliases
- 1110053A20Rik, D2Ertd391e, Harbi1
- Chromosome Location
- Chr. 2: 91674612 - 91710598 on Build GRCm38
- UniGene ID
- Mm.28492
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145528.3 |
NP_663503.1
|
3, 4 |
504 |
|
GenBank mRNA |
AK149672.1 |
- |
3, 4 |
463 |
|
AK154470.1 |
- |
3, 4 |
490 |
|
AK163962.1 |
- |
3, 4 |
456 |
|
AK171206.1 |
- |
3, 4 |
490 |
|
AK171440.1 |
- |
3, 4 |
481 |
|
BC016669.1 |
- |
3, 4 |
488 |
2648797 |
BC023673.1 |
- |
3, 4 |
454 |
5343236 |
BC023712.1 |
- |
3, 4 |
477 |
5346303 |
BC033419.1 |
- |
3, 4 |
455 |
5359944 |
View Full Product Details