Gene Transcripts
- Gene Symbol
- Mta2
- Entrez Gene ID
- 23942
- Gene Name
- metastasis-associated gene family, member 2
- Gene Aliases
- AW550797, Mata1l1, Mta1l1, mmta2
- Chromosome Location
- Chr. 19: 8941920 - 8952300 on Build GRCm38
- UniGene ID
- Mm.25339
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011842.3 |
NP_035972.3
|
5, 6 |
815 |
|
GenBank mRNA |
AF159259.1 |
- |
5, 6 |
690 |
|
AK077548.1 |
- |
5, 6 |
797 |
|
AK088158.1 |
- |
5, 6 |
728 |
|
AK147099.1 |
- |
5, 6 |
794 |
|
AK149834.1 |
- |
5, 6 |
861 |
|
AK157717.1 |
- |
5, 6 |
719 |
|
AK159979.1 |
- |
5, 6 |
791 |
|
AK180159.1 |
- |
3, 4 |
242 |
|
AK185749.1 |
- |
3, 4 |
226 |
|
AK206505.1 |
- |
2, 3 |
212 |
|
BC079847.1 |
- |
5, 6 |
815 |
6419411 |
View Full Product Details