Gene Transcripts

Gene Symbol
Asrgl1
Entrez Gene ID
246307
Gene Name
asparaginase like 1
Gene Aliases
Gliap, Hiob2
Chromosome Location
Chr. 1: 225309737 - 225329743 on Build Rnor_6.0
UniGene ID
Rn.22774
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145089.4 NP_659557.1 2 274
GenBank mRNA AF329099.1 - 2 274
AJ427914.2 - 288
FQ212762.1 - 2 248

View Full Product Details