Gene Transcripts

Gene Symbol
Snx16
Entrez Gene ID
64088
Gene Name
sorting nexin 16
Gene Aliases
Chromosome Location
Chr. 2: 93518796 - 93541076 on Build Rnor_6.0
UniGene ID
Rn.64585
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022289.3 NP_071625.2 2 455
XM_006232135.2 XP_006232197.1 2 468
GenBank mRNA BC061554.1 - 2 459

View Full Product Details