Gene Transcripts
- Gene Symbol
- Gpr64
- Entrez Gene ID
- 266735
- Gene Name
- G protein-coupled receptor 64
- Gene Aliases
- He6, Re6
- Chromosome Location
- Chr. X: 36930186 - 37054925 on Build Rnor_6.0
- UniGene ID
- Rn.57243
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181366.2 |
NP_852031.1
|
3, 4 |
205 |
|
GenBank mRNA |
AF538953.1 |
- |
3, 4 |
215 |
|
View Full Product Details