Gene Transcripts

Gene Symbol
Sp7
Entrez Gene ID
300260
Gene Name
Sp7 transcription factor
Gene Aliases
Osx
Chromosome Location
Chr. 7: 143958520 - 143967484 on Build Rnor_6.0
UniGene ID
Rn.92200
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037632.1 NP_001032721.1 2 364
NM_181374.2 NP_852039.2 2 244
XM_006242390.2 XP_006242452.1 2 600
XM_008765706.1 XP_008763928.1 3 170
GenBank mRNA AY177399.1 - 2 146

View Full Product Details