Gene Transcripts

Gene Symbol
Avpr1b
Entrez Gene ID
29462
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
Chromosome Location
Chr. 13: 48367307 - 48379134 on Build Rnor_6.0
UniGene ID
Rn.10096
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017205.3 NP_058901.3 2 812
XM_008769445.1 XP_008767667.1 1 472
GenBank mRNA AF064541.1 - 1 182
D45400.1 - 1 329
U27322.1 - 1 705

Gene Symbol
LOC100909648
Entrez Gene ID
100909648
Gene Name
vasopressin V1b receptor-like
Gene Aliases
Chromosome Location
Chr. 13: 48390434 - 48400768 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289800.1 NP_001276729.1 1 280
XM_008769404.1 XP_008767626.1 1 465

View Full Product Details