Gene Transcripts

Gene Symbol
Atp1a4
Entrez Gene ID
29132
Gene Name
ATPase, Na+/K+ transporting, alpha 4 polypeptide
Gene Aliases
Chromosome Location
Chr. 13: 90605794 - 90641772 on Build Rnor_6.0
UniGene ID
Rn.224462
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271030.1 NP_001257959.1 13 1813
NM_022848.3 NP_074039.1 13 1816
GenBank mRNA BC087015.1 - 13 1805
U15176.1 - 13 1816

View Full Product Details