Gene Transcripts
- Gene Symbol
- Ybx1
- Entrez Gene ID
- 500538
- Gene Name
- Y box binding protein 1
- Gene Aliases
- Byb1, Cbfa, Dbpb, Ef1a, Msy1, Nsep1, Yb1
- Chromosome Location
- Chr. 5: 138319753 - 138336721 on Build Rnor_6.0
- UniGene ID
- Rn.110976
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031563.3 |
NP_113751.3
|
2, 3 |
355 |
|
XM_008764058.1 |
XP_008762280.1
|
2, 3 |
395 |
|
XM_008764059.1 |
XP_008762281.1
|
2, 3 |
595 |
|
GenBank mRNA |
AB046928.2 |
- |
2, 3 |
514 |
|
BC072486.1 |
- |
2, 3 |
355 |
7105037 |
BC098672.1 |
- |
2, 3 |
355 |
7124218 |
BC103637.1 |
- |
2, 3 |
349 |
7107271 |
D13309.1 |
- |
2, 3 |
325 |
|
M57299.1 |
- |
2, 3 |
331 |
|
M69138.1 |
- |
2, 3 |
491 |
|
View Full Product Details