Gene Transcripts

Gene Symbol
Psmb6
Entrez Gene ID
29666
Gene Name
proteasome (prosome, macropain) subunit, beta type 6
Gene Aliases
Psmb6l
Chromosome Location
Chr. 10: 57131347 - 57133685 on Build Rnor_6.0
UniGene ID
Rn.8118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057099.3 NP_476440.2 6 755
GenBank mRNA BC058451.1 - 6 759
D10754.1 - 6 744

View Full Product Details