Gene Transcripts

Gene Symbol
Mapk10
Entrez Gene ID
25272
Gene Name
mitogen activated protein kinase 10
Gene Aliases
Jnk3, SAPKC, SAPb, Serk2
Chromosome Location
Chr. 14: 8080274 - 8372086 on Build Rnor_6.0
UniGene ID
Rn.9911
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270556.1 NP_001257485.1 14 2087
NM_012806.2 NP_036938.2 14 2114
XM_008769992.1 XP_008768214.1 13 4906
GenBank mRNA L27128.1 - 14 1812

View Full Product Details