Gene Transcripts
- Gene Symbol
- Nsg1
- Entrez Gene ID
- 25247
- Gene Name
- neuron specific gene family member 1
- Gene Aliases
- Bsmrb, Neep21, PEP1
- Chromosome Location
- Chr. 14: 77380264 - 77402007 on Build Rnor_6.0
- UniGene ID
- Rn.2865
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024128.3 |
NP_077042.2
|
2, 3 |
213 |
|
GenBank mRNA |
BC060538.1 |
- |
2, 3 |
216 |
|
View Full Product Details