Gene Transcripts

Gene Symbol
Obp1f
Entrez Gene ID
192267
Gene Name
odorant binding protein I f
Gene Aliases
Obp-1f
Chromosome Location
Chr. X: 45271742 - 45284357 on Build Rnor_6.0
UniGene ID
Rn.9718
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138903.1 NP_620258.1 181
GenBank mRNA AJ251322.1 - 165
J03093.1 - 181

Gene Symbol
LOC100362409
Entrez Gene ID
100362409
Gene Name
odorant-binding protein-like
Gene Aliases
Chromosome Location
Chr. X: 83859898 - 83864401 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257194.2 XP_006257256.1 2 432

Gene Symbol
LOC103690319
Entrez Gene ID
103690319
Gene Name
odorant-binding protein
Gene Aliases
Chromosome Location
Chr. X: 45212961 - 45218173 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773238.1 XP_008771460.1 2 432

View Full Product Details