Gene Transcripts

Gene Symbol
Scgb2a2
Entrez Gene ID
361725
Gene Name
secretoglobin, family 2A, member 2
Gene Aliases
Chromosome Location
Chr. 1: 225899014 - 225902676 on Build Rnor_6.0
UniGene ID
Rn.103180
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203333.2 NP_976078.1 3 648
GenBank mRNA BC062401.1 - 3 608

View Full Product Details