Gene Transcripts
- Gene Symbol
- FAM216A
- Entrez Gene ID
- 29902
- Gene Name
- family with sequence similarity 216 member A
- Gene Aliases
- C12orf24, HSU79274
- Chromosome Location
- Chr. 12: 110468424 - 110490387 on Build GRCh38
- UniGene ID
- Hs.436618
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013300.2 |
NP_037432.2
|
2, 3 |
635 |
|
XM_005253875.4 |
XP_005253932.1
|
2, 3 |
145 |
|
XM_011538246.2 |
XP_011536548.1
|
2, 3 |
163 |
|
GenBank mRNA |
AK225731.1 |
- |
2, 3 |
626 |
|
AK296280.1 |
- |
2, 3 |
647 |
|
AK297684.1 |
- |
2, 3 |
145 |
|
AK308802.1 |
- |
2, 3 |
248 |
|
BC020967.1 |
- |
2, 3 |
451 |
3848037 |
CR457001.1 |
- |
2, 3 |
186 |
|
U79274.1 |
- |
2, 3 |
602 |
|
View Full Product Details