Gene Transcripts

Gene Symbol
MEP1B
Entrez Gene ID
4225
Gene Name
meprin A subunit beta
Gene Aliases
Chromosome Location
Chr. 18: 32190024 - 32222607 on Build GRCh38
UniGene ID
Hs.194777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308171.1 NP_001295100.1 11, 12 1629
NM_005925.2 NP_005916.2 11, 12 1629
XM_011526013.2 XP_011524315.1 10, 11 1395
XM_011526014.2 XP_011524316.1 9, 10 1242
GenBank mRNA AY695931.1 - 11, 12 1640
BC136559.1 - 11, 12 1607 9020549
BC144244.1 - 11, 12 1607 9052764
X81333.1 - 11, 12 1628

View Full Product Details