Gene Transcripts

Gene Symbol
MAST3
Entrez Gene ID
23031
Gene Name
microtubule associated serine/threonine kinase 3
Gene Aliases
Chromosome Location
Chr. 19: 18097764 - 18151692 on Build GRCh38
UniGene ID
Hs.466184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015016.1 NP_055831.1 27 4958
XM_005259823.3 XP_005259880.1 27 5349
XM_005259824.2 XP_005259881.1 28 5035
XM_005259825.2 XP_005259882.1 28 5032
XM_005259826.3 XP_005259883.1 26 5145
XM_006722698.2 XP_006722761.1 27 5027
XM_006722699.2 XP_006722762.2 27 5336
XM_006722700.3 XP_006722763.1 26 5118
XM_011527823.1 XP_011526125.1 29 5125
XM_017026506.1 XP_016881995.1 27 5333
XM_017026507.1 XP_016881996.1 26 5315
XM_017026508.1 XP_016881997.1 26 5313
XM_017026509.1 XP_016881998.1 27 5309
XM_017026511.1 XP_016882000.1 28 5423
XM_017026512.1 XP_016882001.1 27 5014
XM_017026513.1 XP_016882002.1 27 5011
XM_017026514.1 XP_016882003.1 28 5018
XM_017026515.1 XP_016882004.1 27 5024
XM_017026516.1 XP_016882005.1 27 5306
GenBank mRNA AB011133.1 - 27 4957

View Full Product Details