Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009128.2 | NP_033154.2 | 6 | 2192 | |
GenBank mRNA | AK083085.1 | - | 6 | 1778 | |
AK083922.1 | - | 6 | 1786 | ||
AK147406.1 | - | 6 | 2195 | ||
BC040384.1 | - | 6 | 1767 | 3155869 | |
M26270.1 | - | 6 | 1780 |