Gene Transcripts

Gene Symbol
LLPH
Entrez Gene ID
84298
Gene Name
LLP homolog, long-term synaptic facilitation
Gene Aliases
C12orf31, hLLP
Chromosome Location
Chr. 12: 66123069 - 66130753 on Build GRCh38
UniGene ID
Hs.504820
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032338.3 NP_115714.1 2 75
GenBank mRNA AK091568.1 - 2 75
BC006002.1 - 2 71 4247279
BC070232.1 - 2 67 4594274
BC107780.1 - 2 76 6698895

View Full Product Details