Gene Transcripts

Gene Symbol
C20orf85
Entrez Gene ID
128602
Gene Name
chromosome 20 open reading frame 85
Gene Aliases
LLC1, bA196N14.1
Chromosome Location
Chr. 20: 58150927 - 58161130 on Build GRCh38
UniGene ID
Hs.43977
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178456.2 NP_848551.1 3 232
GenBank mRNA BC035405.1 - 3 221
BX093883.1 - 3 232

View Full Product Details