Gene Transcripts

Gene Symbol
Fam203a
Entrez Gene ID
315094
Gene Name
family with sequence similarity 203, member A
Gene Aliases
Brp16, Hgh1
Chromosome Location
Chr. 7: 117434419 - 117437208 on Build Rnor_6.0
UniGene ID
Rn.17102
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007707.1 NP_001007708.1 6 1834
GenBank mRNA BC079158.1 - 6 1834

View Full Product Details