Gene Transcripts

Gene Symbol
Cdsn
Entrez Gene ID
386463
Gene Name
corneodesmosin
Gene Aliases
AI747712
Chromosome Location
Chr. 17: 35543082 - 35557180 on Build GRCm38
UniGene ID
Mm.426664
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008424.2 NP_001008424.2 2 1514
GenBank mRNA AK133029.1 - 1 782
BC055373.1 - 2 1511 30058959

View Full Product Details