Gene Transcripts

Gene Symbol
RT1-M6-1
Entrez Gene ID
414785
Gene Name
RT1 class I, locus M6, gene 1
Gene Aliases
Chromosome Location
Chr. 20: 2057877 - 2060794 on Build Rnor_6.0
UniGene ID
Rn.142703
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008852.1 NP_001008852.1 2 138
XM_008772698.1 XP_008770920.1 2 139
GenBank mRNA EU091353.1 - 1 53
EU106176.1 - 1 53

View Full Product Details