Gene Transcripts

Gene Symbol
C16orf87
Entrez Gene ID
388272
Gene Name
chromosome 16 open reading frame 87
Gene Aliases
Chromosome Location
Chr. 16: 46802047 - 46831162 on Build GRCh38
UniGene ID
Hs.406551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001436.2 NP_001001436.1 4 585
GenBank mRNA BC056676.1 - 4 589
BX647945.1 - 3 402

View Full Product Details