Gene Transcripts

Gene Symbol
Prss48
Entrez Gene ID
368202
Gene Name
protease, serine 48
Gene Aliases
Esspl, Gm1019
Chromosome Location
Chr. 3: 85973843 - 86006002 on Build GRCm38
UniGene ID
Mm.253822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001650.1 NP_001001650.1 4 324
GenBank mRNA BC116378.1 - 4 324
BC116379.1 - 4 324 40101289

View Full Product Details