Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_212510.1 | NP_997675.1 | 5 | 561 | |
GenBank mRNA | BC058498.1 | - | 5 | 561 | |
FQ209874.1 | - | 4 | 402 | ||
FQ209881.1 | - | 5 | 555 | ||
FQ209956.1 | - | 5 | 561 | ||
FQ216877.1 | - | 5 | 554 | ||
FQ221165.1 | - | 5 | 556 | ||
FQ221243.1 | - | 5 | 555 | ||
FQ221413.1 | - | 5 | 555 | ||
FQ221458.1 | - | 5 | 555 | ||
FQ221831.1 | - | 5 | 562 | ||
FQ222162.1 | - | 5 | 555 | ||
FQ222289.1 | - | 5 | 555 | ||
FQ222457.1 | - | 5 | 555 | ||
FQ222662.1 | - | 5 | 555 | ||
FQ223818.1 | - | 5 | 556 | ||
FQ228440.1 | - | 5 | 555 | ||
X14181.1 | - | 5 | 555 |