Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300728.1 | NP_001287657.1 | 4 | 3006 | |
NM_015251.2 | NP_056066.2 | 4 | 3066 | ||
GenBank mRNA | AB007891.1 | - | 3 | 3542 | |
AK025058.1 | - | 1 | 903 | ||
CR749457.1 | - | 4 | 2958 |